Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Actr10
Entrez Gene ID
56444
Gene Name
ARP10 actin-related protein 10
Gene Aliases
Actr11, Arp11
Chromosome Location
Chr.12: 70937857 - 70964718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019785.2 NP_062759.2 4 - 5 438 76
GenBank mRNA AF190797.1 - 4 - 5 405 76
AK090043.1 - 4 - 5 442 76
AK161642.1 - 4 - 5 398 76
BC002227.1 - 4 - 5 398 76

View Full Product Details