Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Herpud2
Entrez Gene ID
80517
Gene Name
HERPUD family member 2
Gene Aliases
5031400M07Rik, AB041580
Chromosome Location
Chr.9: 25108124 - 25151781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020586.2 NP_065611.1 5 - 6 1434 69
GenBank mRNA AB041580.1 - 5 - 6 1229 69
AK019862.1 - 5 - 6 1379 69
AK135935.1 - 5 - 6 1435 69
AK178783.1 - 2 - 3 191 69
AK187953.1 - 2 - 3 195 69
AK214420.1 - 2 - 3 226 69
BC029691.1 - 5 - 6 652 69
BC043693.1 - 5 - 6 1473 69

View Full Product Details