The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020622.2 | NP_065647.1 | 2 - 3 | 258 | 101 |
GenBank mRNA | AF360358.1 | - | 2 - 3 | 259 | 101 |
AF494380.1 | - | 2 - 3 | 163 | 101 | |
AK018571.1 | - | 2 - 3 | 228 | 101 | |
BC048949.1 | - | 2 - 3 | 205 | 101 |