The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042491.2 | NP_001035956.1 | 5 - 6 | 809 | 121 |
NM_001289517.1 | NP_001276446.1 | 5 - 6 | 809 | 121 | |
NM_001289518.1 | NP_001276447.1 | 5 - 6 | 809 | 121 | |
NM_001289519.1 | NP_001276448.1 | 5 - 6 | 809 | 121 | |
NM_001289520.1 | NP_001276449.1 | 5 - 6 | 809 | 121 | |
NM_021505.3 | NP_067480.2 | 5 - 6 | 809 | 121 | |
GenBank mRNA | AB041593.1 | - | 5 - 6 | 752 | 121 |
AK003821.1 | - | 2 - 3 | 202 | 121 | |
AK031432.1 | - | 5 - 6 | 734 | 121 | |
AK049719.1 | - | 5 - 6 | 763 | 121 | |
AK075969.1 | - | 5 - 6 | 748 | 121 | |
AK088327.1 | - | 5 - 6 | 772 | 121 | |
AK147951.1 | - | 5 - 6 | 748 | 121 | |
AK148679.1 | - | 5 - 6 | 764 | 121 | |
AK151314.1 | - | 5 - 6 | 755 | 121 | |
AK151905.1 | - | 5 - 6 | 755 | 121 | |
AK152528.1 | - | 5 - 6 | 725 | 121 | |
AK152981.1 | - | 5 - 6 | 755 | 121 | |
AK153373.1 | - | 5 - 6 | 755 | 121 | |
AK153422.1 | - | 5 - 6 | 755 | 121 | |
AK159204.1 | - | 5 - 6 | 755 | 121 | |
AK159713.1 | - | 5 - 6 | 748 | 121 | |
AK159916.1 | - | 5 - 6 | 748 | 121 | |
AK167561.1 | - | 5 - 6 | 748 | 121 | |
AK168135.1 | - | 5 - 6 | 733 | 121 | |
AK168737.1 | - | 5 - 6 | 749 | 121 | |
AK168814.1 | - | 5 - 6 | 748 | 121 | |
AK169196.1 | - | 5 - 6 | 772 | 121 | |
BC010339.1 | - | 5 - 6 | 725 | 121 | |
BC046566.1 | - | 5 - 6 | 788 | 121 | |
BC046804.1 | - | 5 - 6 | 746 | 121 |