The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024436.3 | NP_077756.2 | 3 - 4 | 462 | 68 |
GenBank mRNA | AB232616.1 | - | 3 - 4 | 201 | 68 |
AJ311124.1 | - | 3 - 4 | 258 | 68 | |
AK014407.1 | - | - | 448 | 68 | |
AK031667.1 | - | 3 - 4 | 442 | 68 | |
AK146594.1 | - | 3 - 4 | 462 | 68 | |
AK149200.1 | - | - | 454 | 68 | |
BC006596.1 | - | 3 - 4 | 449 | 68 |