Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alkbh7
Entrez Gene ID
66400
Gene Name
alkB, alkylation repair homolog 7 (E. coli)
Gene Aliases
2310045B01Rik, 2510008E23Rik, Abh7, Spata11
Chromosome Location
Chr.17: 56997339 - 56999340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025538.3 NP_079814.1 1 - 2 231 76
GenBank mRNA AK007380.1 - 1 - 2 209 76
AK009812.1 - 1 - 2 218 76
BC029677.1 - 1 - 2 198 76

View Full Product Details