The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025538.3 | NP_079814.1 | 3 - 4 | 529 | 99 |
NM_027372.1 | NP_081648.1 | 2 - 3 | 355 | 99 | |
GenBank mRNA | AK009812.1 | - | 3 - 4 | 516 | 99 |
AK010930.1 | - | 2 - 3 | 356 | 99 | |
BC029677.1 | - | 3 - 4 | 496 | 99 |