The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025671.2 | NP_079947.1 | 2 - 3 | 188 | 86 |
GenBank mRNA | AK004943.1 | - | 2 - 3 | 189 | 86 |
AK017497.1 | - | 2 - 3 | 189 | 86 | |
AK018534.1 | - | 2 - 3 | 189 | 86 | |
AK168946.1 | - | 2 - 3 | 197 | 86 | |
BC022762.1 | - | 1 - 2 | 36 | 86 | |
BC069872.1 | - | 2 - 3 | 191 | 86 |