The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025689.4 | NP_079965.2 | 2 - 3 | 385 | 55 |
GenBank mRNA | AK014056.1 | - | 1 - 2 | 325 | 55 |
AK017860.1 | - | 2 - 3 | 382 | 55 | |
AK137013.1 | - | 2 - 3 | 389 | 55 | |
AK141240.1 | - | 2 - 3 | 388 | 55 | |
BC098221.1 | - | 1 - 2 | 331 | 55 | |
BC116786.1 | - | 1 - 2 | 243 | 55 | |
BC116788.1 | - | 1 - 2 | 243 | 55 |