The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037726.1 | NP_001032815.1 | 7 - 7 | 3919 | 79 |
NM_009952.2 | NP_034082.1 | 9 - 9 | 4083 | 79 | |
NM_133828.2 | NP_598589.2 | 8 - 8 | 4041 | 79 | |
GenBank mRNA | AK014391.1 | - | 1 - 1 | 552 | 79 |
AK032372.1 | - | 1 - 1 | 534 | 79 | |
BC048151.1 | - | 1 - 1 | 307 | 79 | |
BC058366.1 | - | 1 - 1 | 307 | 79 | |
BC071201.1 | - | 1 - 1 | 739 | 79 |