Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl28
Entrez Gene ID
66689
Gene Name
kelch-like 28
Gene Aliases
2810440N09Rik, 4122402F11Rik, 4931401E10Rik, Btbd5
Chromosome Location
Chr.12: 64942198 - 64965605 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025707.3 NP_079983.1 4 - 5 1682 85
GenBank mRNA AK002637.1 - 4 - 5 1649 85
AK012778.2 - 4 - 5 1662 85
AK014466.1 - 4 - 5 1683 85
AK016422.1 - 4 - 5 1624 85
BC018469.1 - 4 - 5 1638 85
BC037017.1 - 4 - 5 1655 85

View Full Product Details