Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx31
Entrez Gene ID
66696
Gene Name
sorting nexin 31
Gene Aliases
4631426E05Rik
Chromosome Location
Chr.15: 36504062 - 36555614 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025712.4 NP_079988.3 10 - 11 1034 101
GenBank mRNA AK014536.1 - 10 - 11 1036 101
AK034196.1 - 10 - 11 1036 101
AK035226.1 - 10 - 11 1035 101
BC061007.1 - 10 - 11 1030 101

View Full Product Details