The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025833.3 | NP_080109.1 | 12 - 13 | 1707 | 62 |
GenBank mRNA | AK004918.2 | - | 12 - 13 | 1708 | 62 |
AK008203.1 | - | 12 - 13 | 1608 | 62 | |
AK162158.1 | - | - | 1573 | 62 | |
BC015459.1 | - | 12 - 13 | 1485 | 62 |