The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025873.2 | NP_080149.2 | 1 - 2 | 184 | 75 |
GenBank mRNA | AK003556.1 | - | 1 - 2 | 185 | 75 |
AK010176.1 | - | 1 - 2 | 184 | 75 | |
AK128921.1 | - | 1 - 2 | 183 | 75 | |
BC019812.1 | - | 1 - 2 | 153 | 75 | |
BC051040.1 | - | 1 - 2 | 160 | 75 |