Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plin5
Entrez Gene ID
66968
Gene Name
perilipin 5
Gene Aliases
2310076L09Rik, AI415325, AW109675, Lsdp5, MLDP, PAT-1
Chromosome Location
Chr.17: 56111597 - 56117606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077348.1 NP_001070816.1 4 - 5 562 61
NM_025874.3 NP_080150.2 5 - 6 508 61
GenBank mRNA AK010202.1 - 5 - 6 508 61
AK075872.1 - 5 - 6 505 61
AY919875.1 - 4 - 5 521 61
BC024138.1 - 5 - 6 488 61
DQ473305.1 - 5 - 6 488 61

View Full Product Details