The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025942.2 | NP_080218.1 | 6 - 7 | 787 | 128 |
NM_030091.1 | NP_084367.1 | 6 - 7 | 787 | 128 | |
GenBank mRNA | AK010817.1 | - | 6 - 7 | 703 | 128 |
AK013055.1 | - | 6 - 7 | 788 | 128 | |
AK019142.1 | - | 6 - 7 | 673 | 128 | |
AK159873.1 | - | 6 - 7 | 704 | 128 | |
AK167458.1 | - | 6 - 7 | 703 | 128 | |
AK170982.1 | - | 6 - 7 | 717 | 128 | |
BC011318.1 | - | 6 - 7 | 687 | 128 | |
BC029207.1 | - | 6 - 7 | 680 | 128 | |
BG069761.2 | - | 2 - 3 | 128 |