Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bfar
Entrez Gene ID
67118
Gene Name
bifunctional apoptosis regulator
Gene Aliases
3010001A07Rik, AI666707, AW107665, Bar, Rnf47
Chromosome Location
Chr.16: 13671858 - 13703612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177552.1 NP_001171023.1 1 - 2 217 65
NM_025976.5 NP_080252.1 1 - 2 217 65
GenBank mRNA AK013874.1 - 1 - 2 82 65
AK028592.1 - 1 - 2 56 65
AK041839.1 - 1 - 2 126 65
BC027221.1 - 1 - 2 65
BY329099.1 - 1 - 2 218 65

View Full Product Details