The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026229.4 | NP_080505.1 | 7 - 8 | 694 | 74 |
GenBank mRNA | AK009734.1 | - | 6 - 7 | 573 | 74 |
AK016787.1 | - | 7 - 8 | 694 | 74 | |
AK034931.1 | - | - | 262 | 74 | |
BC010729.1 | - | 7 - 8 | 687 | 74 |