The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026240.2 | NP_080516.2 | 6 - 7 | 729 | 69 |
GenBank mRNA | AK018551.1 | - | 7 - 8 | 979 | 69 |
AK018691.1 | - | 6 - 7 | 715 | 69 | |
AK038289.1 | - | 5 - 6 | 770 | 69 | |
AK161329.1 | - | 6 - 7 | 662 | 69 | |
AK164050.1 | - | 6 - 7 | 714 | 69 | |
BC057978.1 | - | 6 - 7 | 704 | 69 |