Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gin1
Entrez Gene ID
252876
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2
Chromosome Location
Chr.1: 97770168 - 97793710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026250.2 NP_080526.2 3 - 4 418 62
GenBank mRNA AK014287.1 - 3 - 4 430 62
AK015243.1 - 3 - 4 397 62
AK169361.1 - 3 - 4 381 62
BC033572.1 - 2 - 3 363 62

View Full Product Details