The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026251.2 | NP_080527.2 | 6 - 7 | 784 | 59 |
GenBank mRNA | AK015195.1 | - | 6 - 7 | 785 | 59 |
BC145646.1 | - | 4 - 5 | 562 | 59 | |
DV658440.1 | - | 4 - 5 | 395 | 59 |