Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spaca1
Entrez Gene ID
67652
Gene Name
sperm acrosome associated 1
Gene Aliases
1700124L11Rik, 4930540L03Rik
Chromosome Location
Chr.4: 34024872 - 34050067 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290443.1 NP_001277372.1 2 - 3 362 112
NM_026293.3 NP_080569.1 2 - 3 364 112
GenBank mRNA AK006187.1 - 2 - 3 365 112
AK007270.1 - 2 - 3 336 112
AK016011.1 - 2 - 3 357 112
BC147079.1 - 2 - 3 361 112
BC147080.1 - 2 - 3 361 112
BY706285.1 - 2 - 3 363 112

View Full Product Details