The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026323.2 | NP_080599.1 | 4 - 5 | 532 | 94 |
GenBank mRNA | AF334269.1 | - | 4 - 5 | 529 | 94 |
AK003771.1 | - | 2 - 3 | 175 | 94 | |
AK005519.1 | - | 4 - 5 | 533 | 94 | |
BC099427.1 | - | 4 - 5 | 523 | 94 |