The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026380.3 | NP_080656.2 | 7 - 7 | 4461 | 68 |
GenBank mRNA | AK018215.1 | - | 1 - 1 | 254 | 68 |
AK018337.1 | - | 1 - 1 | 446 | 68 | |
BC056441.1 | - | 1 - 1 | 1467 | 68 | |
BC057559.1 | - | 1 - 1 | 893 | 68 | |
BC065056.1 | - | 7 - 7 | 4394 | 68 | |
BC094577.1 | - | 6 - 6 | 4077 | 68 |