The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026558.4 | NP_080834.3 | 2 - 3 | 552 | 104 |
GenBank mRNA | AK019574.1 | - | 2 - 3 | 380 | 104 |
AK078438.1 | - | 2 - 3 | 533 | 104 | |
AK088705.1 | - | 2 - 3 | 533 | 104 | |
BC020162.1 | - | - | 346 | 104 |