The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028021.1 | 2 - 3 | 309 | 88 | |
GenBank mRNA | AK015792.1 | - | 2 - 3 | 310 | 88 |
AK019686.1 | - | 2 - 3 | 310 | 88 |