Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc21
Entrez Gene ID
68268
Gene Name
zinc finger, DHHC domain containing 21
Gene Aliases
9130404H11Rik, AL024349, D130004H04Rik, dep
Chromosome Location
Chr.4: 82798738 - 82859891 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026647.3 NP_080923.2 7 - 8 977 124
GenBank mRNA AK020293.1 - 7 - 8 859 124
AY166674.1 - 5 - 6 615 124
BC043110.1 - 7 - 8 823 124
BC058372.1 - 7 - 8 845 124

View Full Product Details