The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026947.4 | NP_081223.1 | 1 - 2 | 52 | 85 |
GenBank mRNA | AK007578.1 | - | 1 - 2 | 53 | 85 |
AK131590.1 | - | 1 - 2 | 53 | 85 |