The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027036.3 | NP_081312.2 | 1 - 1 | 561 | 81 |
GenBank mRNA | AK005588.1 | - | 1 - 1 | 560 | 81 |
BC048744.1 | - | 2 - 2 | 521 | 81 | |
BC061030.1 | - | 1 - 1 | 557 | 81 | |
BC087910.1 | - | 2 - 2 | 447 | 81 | |
BC087934.1 | - | 2 - 2 | 517 | 81 | |
CA465179.1 | - | 1 - 1 | 560 | 81 |