Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spaca4
Entrez Gene ID
69363
Gene Name
sperm acrosome associated 4
Gene Aliases
1700008E09Rik, AV043694, Samp14
Chromosome Location
Chr.7: 45725107 - 45725816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027055.3 NP_081331.2 1 - 1 261 56
GenBank mRNA AK005760.1 - 1 - 1 253 56

View Full Product Details