The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027117.3 | NP_081393.2 | 11 - 12 | 1362 | 87 |
GenBank mRNA | AK009481.1 | - | 11 - 12 | 1277 | 87 |
AY513272.1 | - | 11 - 12 | 1313 | 87 | |
BC005581.1 | - | 11 - 12 | 1286 | 87 |