The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027294.2 | NP_081570.1 | 3 - 4 | 664 | 75 |
GenBank mRNA | AK012262.1 | - | 3 - 4 | 665 | 75 |
AY243513.1 | - | 3 - 4 | 699 | 75 | |
BC027554.1 | - | 3 - 4 | 586 | 75 |