The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027360.2 | NP_081636.1 | 3 - 4 | 203 | 135 |
GenBank mRNA | AK008333.1 | - | 3 - 4 | 204 | 135 |
BC055712.1 | - | 3 - 4 | 182 | 135 | |
BC059719.1 | - | 3 - 4 | 191 | 135 |