Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mier2
Entrez Gene ID
70427
Gene Name
mesoderm induction early response 1, family member 2
Gene Aliases
2700087H15Rik, mKIAA1193
Chromosome Location
Chr.10: 79540245 - 79555631 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027422.2 NP_081698.2 13 - 14 1603 100
GenBank mRNA AK012576.1 - 1 - 2 452 100
AK154672.1 - 13 - 14 1620 100
AK173128.1 - 12 - 13 1758 100
BC067013.1 - 13 - 14 1603 100
BC080757.1 - 6 - 7 854 100
BC083321.1 - 4 - 5 724 100

View Full Product Details