Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zcchc8
Entrez Gene ID
70650
Gene Name
zinc finger, CCHC domain containing 8
Gene Aliases
5730565F05Rik
Chromosome Location
Chr.5: 123698302 - 123721044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027494.3 NP_081770.3 6 - 7 726 64
GenBank mRNA AK017857.1 - 6 - 7 622 64
AK032349.1 - 6 - 7 703 64
AK156770.1 - 6 - 7 698 64
AK166990.1 - 6 - 7 633 64
AK167126.1 - 6 - 7 631 64
AK167736.1 - 6 - 7 633 64
BC013555.1 - 5 - 6 378 64

View Full Product Details