Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmrtc1a
Entrez Gene ID
70887
Gene Name
DMRT-like family C1a
Gene Aliases
1700030H14Rik, 4921520P21Rik, DMRTC1, Dmrt8.1
Chromosome Location
Chr.X: 102903216 - 102934003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038616.2 NP_001033705.2 1 - 2 191 75
NM_027591.3 NP_081867.1 3 - 4 289 75
NM_029378.2 NP_083654.1 1 - 2 103 75
GenBank mRNA AK014934.1 - 3 - 4 290 75
BC147175.1 - 3 - 4 261 75
BC147176.1 - 3 - 4 261 75
DQ316772.1 - 3 - 4 245 75
DQ316773.1 - 1 - 2 103 75

View Full Product Details