Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggt6
Entrez Gene ID
71522
Gene Name
gamma-glutamyltransferase 6
Gene Aliases
9030405D14Rik, AI427679, AW490214
Chromosome Location
Chr.11: 72435243 - 72438566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027819.2 NP_082095.2 2 - 3 446 69
GenBank mRNA AK018485.1 - 2 - 3 448 69
BC058747.1 - 2 - 3 446 69

View Full Product Details