The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027829.3 | NP_082105.3 | 2 - 3 | 352 | 65 |
GenBank mRNA | AK018541.1 | - | 2 - 3 | 134 | 65 |
BC057953.1 | - | 2 - 3 | 278 | 65 | |
BF682661.1 | - | 2 - 3 | 287 | 65 | |
CB055593.1 | - | 2 - 3 | 256 | 65 |