The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027901.2 | NP_082177.2 | 4 - 5 | 1133 | 68 |
GenBank mRNA | AK004895.1 | - | 4 - 5 | 911 | 68 |
AK035792.1 | - | 4 - 5 | 899 | 68 | |
AK043944.1 | - | 4 - 5 | 900 | 68 | |
AK083585.1 | - | 4 - 5 | 1134 | 68 | |
AK083636.1 | - | 3 - 4 | 888 | 68 | |
AK129033.1 | - | 3 - 4 | 883 | 68 | |
AK138760.1 | - | - | 902 | 68 | |
AK149771.1 | - | 5 - 6 | 1030 | 68 | |
AK151300.1 | - | 4 - 5 | 898 | 68 | |
AK152439.1 | - | 4 - 5 | 898 | 68 | |
AK154056.1 | - | 4 - 5 | 895 | 68 | |
AK159938.1 | - | 4 - 5 | 900 | 68 | |
BC034369.1 | - | 4 - 5 | 911 | 68 | |
BC043100.1 | - | 4 - 5 | 1121 | 68 |