The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028018.2 | NP_082294.1 | 4 - 5 | 488 | 86 |
GenBank mRNA | AK010262.1 | - | 4 - 5 | 458 | 86 |
AK050468.1 | - | 4 - 5 | 455 | 86 | |
AK088192.1 | - | 4 - 5 | 443 | 86 | |
AK122215.1 | - | 2 - 3 | 262 | 86 | |
AK152830.1 | - | 4 - 5 | 455 | 86 | |
AK161899.1 | - | 4 - 5 | 444 | 86 | |
AK207302.1 | - | 2 - 3 | 182 | 86 | |
AK218302.1 | - | 2 - 3 | 120 | 86 | |
BC017605.1 | - | 4 - 5 | 454 | 86 | |
BC039052.1 | - | 4 - 5 | 431 | 86 |