Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ist1
Entrez Gene ID
71955
Gene Name
increased sodium tolerance 1 homolog (yeast)
Gene Aliases
2400003C14Rik, AW536298, mKIAA0174
Chromosome Location
Chr.8: 109671321 - 109693294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028018.2 NP_082294.1 8 - 9 978 90
GenBank mRNA AK010262.1 - 8 - 9 948 90
AK088192.1 - 8 - 9 933 90
AK152830.1 - 8 - 9 945 90
AK186831.1 - 2 - 3 224 90
AK215347.1 - 2 - 3 208 90
BC017605.1 - 8 - 9 944 90

View Full Product Details