Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx10
Entrez Gene ID
71982
Gene Name
sorting nexin 10
Gene Aliases
2410004M09Rik
Chromosome Location
Chr.6: 51523902 - 51590679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127349.1 NP_001120821.1 1 - 2 132 111
GenBank mRNA AK010399.1 - 1 - 2 75 111
AK150704.1 - 1 - 2 135 111
AK150999.1 - 1 - 2 135 111
AK151320.1 - 1 - 2 135 111
AK151520.1 - 1 - 2 135 111
AK152825.1 - 1 - 2 107 111
AK152949.1 - 1 - 2 123 111
AK153127.1 - 1 - 2 135 111
AK153356.1 - 1 - 2 85 111
AK169110.1 - 1 - 2 66 111
BC010334.1 - 1 - 2 64 111

View Full Product Details