The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114322.1 | NP_001107794.1 | 3 - 4 | 395 | 70 |
NM_028069.3 | NP_082345.1 | 3 - 4 | 395 | 70 | |
GenBank mRNA | AF462391.1 | - | 3 - 4 | 384 | 70 |
AK007988.1 | - | 3 - 4 | 384 | 70 | |
AK027913.1 | - | 3 - 4 | 396 | 70 | |
BB609487.1 | - | 3 - 4 | 396 | 70 | |
BC043336.1 | - | 3 - 4 | 360 | 70 | |
BC054469.1 | - | 3 - 4 | 380 | 70 |