Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sulf2
Entrez Gene ID
72043
Gene Name
sulfatase 2
Gene Aliases
2010004N24Rik, AU020235, MSulf-2, mKIAA1247
Chromosome Location
Chr.2: 166073899 - 166155683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252578.1 NP_001239507.1 17 - 18 2815 80
NM_001252579.1 NP_001239508.1 17 - 18 2661 80
NM_028072.5 NP_082348.2 17 - 18 2657 80
GenBank mRNA AK008108.1 - 8 - 9 920 80
AK028874.1 - 17 - 18 2572 80
AK034712.1 - 17 - 18 2642 80
AK036685.1 - 17 - 18 2593 80
AK049170.1 - 12 - 13 1572 80
AK081643.1 - 17 - 18 2615 80
AK129316.1 - 17 - 18 2815 80
AK133336.1 - - 2631 80
AK165183.1 - 17 - 18 2569 80
AY101177.1 - 16 - 17 2476 80
BC027238.1 - 6 - 7 631 80
BC062900.1 - 17 - 18 2613 80
BC141086.1 - 16 - 17 2410 80

View Full Product Details