The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177886.1 | NP_001171357.1 | 1 - 2 | 90 | 70 |
NM_001177887.1 | NP_001171358.1 | 1 - 2 | 94 | 70 | |
NM_028078.3 | NP_082354.1 | 1 - 2 | 87 | 70 | |
GenBank mRNA | AK008060.1 | - | 1 - 2 | 58 | 70 |
AK078934.1 | - | 1 - 2 | 58 | 70 | |
AK086973.1 | - | 1 - 2 | 91 | 70 | |
BX511978.1 | - | 1 - 2 | 118 | 70 | |
BY033616.1 | - | 1 - 2 | 88 | 70 |