Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adck1
Entrez Gene ID
72113
Gene Name
aarF domain containing kinase 1
Gene Aliases
2610005A10Rik
Chromosome Location
Chr.12: 88360491 - 88461726 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277296.1 NP_001264225.1 3 - 4 502 70
NM_001277297.1 NP_001264226.1 3 - 4 314 70
NM_028105.4 NP_082381.1 3 - 4 423 70
GenBank mRNA AK011310.1 - 3 - 4 274 70
AK050868.1 - 3 - 4 404 70
AK154839.1 - 3 - 4 369 70
BC010539.1 - 3 - 4 316 70
BY242507.1 - 3 - 4 321 70

View Full Product Details