The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028184.3 | NP_082460.2 | 1 - 2 | 140 | 73 |
GenBank mRNA | AK008702.1 | - | 1 - 2 | 200 | 73 |
AK046447.1 | - | 1 - 2 | 239 | 73 | |
AK083244.1 | - | 1 - 2 | 197 | 73 | |
AK169310.1 | - | 1 - 2 | 164 | 73 | |
BC006906.1 | - | 1 - 2 | 51 | 73 | |
BC013564.1 | - | 1 - 2 | 140 | 73 |