The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029410.3 | NP_083686.1 | 3 - 4 | 374 | 57 |
GenBank mRNA | AK013411.1 | - | 3 - 4 | 348 | 57 |
AK017362.1 | - | 3 - 4 | 375 | 57 | |
BC039807.1 | - | - | 526 | 57 |