The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028584.3 | NP_082860.2 | 1 - 2 | 490 | 79 |
NM_212447.2 | NP_997612.2 | 1 - 2 | 490 | 79 | |
GenBank mRNA | AK007346.1 | - | 1 - 2 | 411 | 79 |
AW107118.1 | - | 2 - 3 | 310 | 79 | |
BB619320.1 | - | 1 - 2 | 79 | ||
BC025851.1 | - | 1 - 2 | 453 | 79 | |
BQ900249.1 | - | 1 - 2 | 131 | 79 |