The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028711.3 | NP_082987.1 | 1 - 2 | 118 | 95 |
GenBank mRNA | AB106930.1 | - | 1 - 2 | 262 | 95 |
AK014394.1 | - | 1 - 2 | 119 | 95 | |
AK021314.1 | - | 2 - 3 | 204 | 95 | |
AK043831.1 | - | 1 - 2 | 318 | 95 | |
BC138993.1 | - | 1 - 2 | 125 | 95 | |
BC138994.1 | - | 1 - 2 | 125 | 95 |